Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100471.4 | NP_563752.1 | 5 - 6 | 575 | 112 | |
NM_202038.4 | NP_973767.1 | 5 - 6 | 582 | 112 | ||
GenBank mRNA | AF410268.1 | - | 5 - 6 | 558 | 112 | |
AK316958.1 | - | 5 - 6 | 575 | 112 | ||
AY143860.1 | - | 5 - 6 | 535 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |