Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SK41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035936.1 | NP_001031013.1 | 13 - 14 | 1448 | 68 | |
NM_001084029.2 | NP_001077498.1 | 12 - 13 | 1558 | 68 | ||
NM_001084030.2 | NP_001077499.1 | 12 - 13 | 1596 | 68 | ||
NM_001331855.1 | NP_001322595.1 | 12 - 13 | 1298 | 68 | ||
NM_001331856.1 | NP_001322594.1 | 8 - 9 | 923 | 68 | ||
NM_001331857.1 | NP_001318967.1 | 13 - 14 | 1635 | 68 | ||
NM_001331858.1 | NP_001322592.1 | 12 - 13 | 1714 | 68 | ||
NM_001331859.1 | NP_001322593.1 | 13 - 14 | 1564 | 68 | ||
NM_100858.5 | NP_172455.1 | 12 - 13 | 1643 | 68 | ||
NM_179296.1 | NP_849627.1 | 13 - 14 | 1411 | 68 | ||
NM_202072.2 | NP_973801.1 | 11 - 12 | 1710 | 68 | ||
GenBank mRNA | AY092987.1 | - | 12 - 13 | 1486 | 68 | |
BT000132.1 | - | 11 - 12 | 1176 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |