Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIF3H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100960.3 | NP_563880.1 | 2 - 3 | 403 | 77 | |
NM_202079.1 | NP_973808.1 | 1 - 2 | 289 | 77 | ||
GenBank mRNA | AY054641.1 | - | 2 - 3 | 237 | 77 | |
AY081546.1 | - | 1 - 2 | 86 | 77 | ||
AY087338.1 | - | 2 - 3 | 305 | 77 | ||
BT000765.1 | - | 2 - 3 | 182 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |