Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101198.4 | NP_172785.1 | 3 - 4 | 571 | 142 | |
NM_202086.1 | NP_973815.1 | 3 - 4 | 504 | 142 | ||
GenBank mRNA | BT000449.1 | - | 3 - 4 | 504 | 142 | |
BT001213.1 | - | 3 - 4 | 461 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |