Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332070.1 | NP_001323307.1 | 4 - 5 | 247 | 61 | |
NM_101226.5 | NP_172813.1 | 6 - 7 | 616 | 61 | ||
NM_202088.2 | NP_973817.1 | 5 - 6 | 513 | 61 | ||
GenBank mRNA | AF091843.1 | - | 6 - 7 | 453 | 61 | |
AY080725.1 | - | 6 - 7 | 490 | 61 | ||
AY114062.1 | - | 5 - 6 | 354 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |