Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102081.4 | NP_564167.1 | 6 - 7 | 1053 | 77 | |
NM_202155.2 | NP_973884.1 | 5 - 6 | 1225 | 77 | ||
GenBank mRNA | AY054505.1 | - | 6 - 7 | 808 | 77 | |
AY058834.1 | - | 6 - 7 | 860 | 77 | ||
AY062838.1 | - | 6 - 7 | 808 | 77 | ||
AY081674.1 | - | 6 - 7 | 761 | 77 | ||
AY087580.1 | - | 6 - 7 | 854 | 77 | ||
U36446.1 | - | 6 - 7 | 809 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |