Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSZ21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123870.4 | NP_001117342.1 | 5 - 6 | 616 | 65 | |
NM_001198145.1 | NP_001185074.1 | 5 - 6 | 798 | 65 | ||
NM_102234.4 | NP_564208.1 | 5 - 6 | 797 | 65 | ||
NM_202172.1 | NP_973901.1 | 4 - 5 | 629 | 65 | ||
GenBank mRNA | AF033587.1 | - | 5 - 6 | 610 | 65 | |
AK317576.1 | - | 5 - 6 | 638 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |