Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036016.2 | NP_001031093.1 | 5 - 6 | 753 | 87 | |
NM_001332707.1 | NP_001323071.1 | 5 - 6 | 990 | 87 | ||
NM_001332708.1 | NP_001323072.1 | 5 - 6 | 1189 | 87 | ||
NM_001332709.1 | NP_001323073.1 | 5 - 6 | 1149 | 87 | ||
NM_102390.3 | NP_173950.1 | 4 - 5 | 1256 | 87 | ||
NM_202184.3 | NP_973913.1 | 5 - 6 | 1000 | 87 | ||
GenBank mRNA | AK117355.1 | - | 4 - 5 | 956 | 87 | |
BT005334.1 | - | 4 - 5 | 846 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |