Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAT32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102541.3 | NP_564294.1 | 3 - 4 | 561 | 77 | |
NM_179384.2 | NP_849715.2 | 2 - 3 | 736 | 77 | ||
NM_202194.3 | NP_973923.1 | 3 - 4 | 649 | 77 | ||
GenBank mRNA | AF361609.1 | - | 3 - 4 | 546 | 77 | |
AF428281.1 | - | 3 - 4 | 546 | 77 | ||
AY078052.1 | - | 2 - 3 | 250 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |