Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAT32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102541.3 | NP_564294.1 | 7 - 8 | 975 | 82 | |
NM_179384.2 | NP_849715.2 | 6 - 7 | 1150 | 82 | ||
NM_202194.3 | NP_973923.1 | 7 - 8 | 1063 | 82 | ||
GenBank mRNA | AF361609.1 | - | 7 - 8 | 960 | 82 | |
AF428281.1 | - | 7 - 8 | 960 | 82 | ||
AY078052.1 | - | 6 - 7 | 664 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |