Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337535.1 | NP_001325772.1 | 2 - 3 | 1659 | 72 | |
NM_001337536.1 | NP_001325771.1 | 3 - 4 | 1559 | 72 | ||
NM_001337537.1 | NP_001325773.1 | 4 - 5 | 1073 | 72 | ||
NM_111333.5 | NP_187112.1 | 3 - 4 | 1173 | 72 | ||
GenBank mRNA | AY070475.1 | - | 3 - 4 | 1046 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |