Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IAA16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111345.3 | NP_187124.1 | 2 - 3 | 950 | 151 | |
GenBank mRNA | AF332400.1 | - | 2 - 3 | 474 | 151 | |
AF375420.1 | - | 2 - 3 | 657 | 151 | ||
AK229447.1 | - | 2 - 3 | 657 | 151 | ||
AY087195.1 | - | 2 - 3 | 661 | 151 | ||
BT000857.1 | - | 2 - 3 | 474 | 151 | ||
U49072.1 | - | 2 - 3 | 640 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |