Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337544.1 | NP_001319473.1 | 11 - 12 | 1565 | 74 | |
NM_001337545.1 | NP_001319474.1 | 11 - 12 | 1443 | 74 | ||
NM_111359.4 | NP_187138.1 | 10 - 11 | 1534 | 74 | ||
NM_202493.3 | NP_974222.1 | 11 - 12 | 1430 | 74 | ||
GenBank mRNA | AY059920.1 | - | 10 - 11 | 1395 | 74 | |
AY072096.1 | - | 10 - 11 | 1401 | 74 | ||
AY096583.1 | - | 10 - 11 | 1287 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |