Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337544.1 | NP_001319473.1 | 8 - 9 | 1147 | 87 | |
NM_001337545.1 | NP_001319474.1 | 8 - 9 | 1025 | 87 | ||
NM_111359.4 | NP_187138.1 | 7 - 8 | 1116 | 87 | ||
NM_202493.3 | NP_974222.1 | 8 - 9 | 1012 | 87 | ||
GenBank mRNA | AY059920.1 | - | 7 - 8 | 977 | 87 | |
AY072096.1 | - | 7 - 8 | 983 | 87 | ||
AY096583.1 | - | 7 - 8 | 869 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |