Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111415.5 | NP_187193.3 | 9 - 10 | 981 | 106 | |
NM_202498.2 | NP_974227.1 | 9 - 10 | 1091 | 106 | ||
GenBank mRNA | AK228046.1 | - | 9 - 10 | 982 | 106 | |
BT006458.1 | - | 9 - 10 | 879 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |