Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLDP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111436.6 | NP_187214.2 | 4 - 5 | 720 | 117 | |
GenBank mRNA | AM182458.1 | - | 4 - 5 | 646 | 117 | |
AY305003.1 | - | 4 - 5 | 593 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |