Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RECQI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337605.1 | NP_001327100.1 | 3 - 4 | 863 | 86 | |
NM_111448.3 | NP_187225.2 | 4 - 5 | 983 | 86 | ||
GenBank mRNA | AJ404470.1 | - | 4 - 5 | 950 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |