Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LON4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111453.1 | NP_566259.1 | 6 - 7 | 754 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |