Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCI2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111463.2 | NP_187240.1 | 1 - 2 | 435 | 72 | |
GenBank mRNA | AF122006.1 | - | 1 - 2 | 120 | 72 | |
AK176253.1 | - | 1 - 2 | 139 | 72 | ||
AK228716.1 | - | 1 - 2 | 143 | 72 | ||
AY084701.1 | - | 1 - 2 | 145 | 72 | ||
BT026006.1 | - | 1 - 2 | 80 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |