Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111477.4 | NP_187254.1 | 11 - 12 | 1194 | 108 | |
GenBank mRNA | AB000799.1 | - | 11 - 12 | 1082 | 108 | |
AY069917.1 | - | 11 - 12 | 1176 | 108 | ||
BT001024.1 | - | 11 - 12 | 1041 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |