Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337653.1 | NP_001326702.1 | 1 - 2 | 955 | 135 | |
NM_111513.4 | NP_187289.1 | 2 - 3 | 697 | 135 | ||
GenBank mRNA | AF487270.1 | - | 2 - 3 | 386 | 135 | |
BT026465.1 | - | 2 - 3 | 386 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |