Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHR11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202897.1 | NP_001189826.1 | 1 - 2 | 288 | 78 | |
NM_001202898.2 | NP_001189827.1 | 1 - 2 | 354 | 78 | ||
NM_111515.5 | NP_187291.2 | 1 - 2 | 354 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |