Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111524.4 | NP_187300.3 | 3 - 4 | 962 | 112 | |
GenBank mRNA | AF039406.1 | - | 3 - 4 | 808 | 112 | |
AY035017.1 | - | 3 - 4 | 822 | 112 | ||
AY059083.1 | - | 3 - 4 | 679 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |