Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111527.3 | NP_566285.1 | 6 - 7 | 1341 | 105 | |
GenBank mRNA | AJ491320.1 | - | 6 - 7 | 1257 | 105 | |
AY059889.1 | - | 6 - 7 | 1182 | 105 | ||
BT002100.1 | - | 6 - 7 | 1137 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |