Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IBR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111561.5 | NP_187337.2 | 2 - 3 | 563 | 85 | |
GenBank mRNA | AY091014.1 | - | 2 - 3 | 326 | 85 | |
BT002365.1 | - | 2 - 3 | 272 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |