Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337693.1 | NP_001326712.1 | 10 - 11 | 1283 | 86 | |
NM_001337694.1 | NP_001326713.1 | 9 - 10 | 1151 | 86 | ||
NM_111596.6 | NP_187372.5 | 9 - 10 | 1093 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |