Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIR12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337711.1 | NP_001327216.1 | 2 | 952 | 133 | |
NM_111618.3 | NP_566306.3 | 1 | 1062 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |