Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPKKK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337756.1 | NP_001326459.1 | 8 - 9 | 929 | 98 | |
NM_001337757.1 | NP_001326460.1 | 4 - 5 | 485 | 98 | ||
NM_111677.4 | NP_187455.1 | 8 - 9 | 965 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |