Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KOB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111689.5 | NP_187467.1 | 1 - 2 | 871 | 70 | |
GenBank mRNA | AF543710.1 | - | 1 - 2 | 393 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |