Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036629.2 | NP_001031706.1 | 12 - 13 | 1974 | 99 | |
NM_118530.5 | NP_194129.1 | 12 - 13 | 2035 | 99 | ||
GenBank mRNA | AF082176.1 | - | 12 - 13 | 1873 | 99 | |
AF344312.1 | - | 12 - 13 | 1586 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |