Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036629.2 | NP_001031706.1 | 9 - 10 | 1279 | 70 | |
NM_118530.5 | NP_194129.1 | 9 - 10 | 1334 | 70 | ||
GenBank mRNA | AF082176.1 | - | 9 - 10 | 1172 | 70 | |
AF344312.1 | - | 9 - 10 | 885 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |