Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118544.4 | NP_567694.2 | 1 - 2 | 783 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |