Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLO13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341655.1 | NP_001328763.1 | 10 - 11 | 1206 | 133 | |
NM_118558.4 | NP_567697.1 | 10 - 11 | 1206 | 133 | ||
GenBank mRNA | AF369574.1 | - | 10 - 11 | 1170 | 133 | |
AK221618.1 | - | 10 - 11 | 1159 | 133 | ||
BT026373.1 | - | 10 - 11 | 1140 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |