Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIPK8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084973.1 | NP_001078442.1 | 5 - 6 | 599 | 112 | |
NM_118573.4 | NP_194171.1 | 5 - 6 | 876 | 112 | ||
GenBank mRNA | BT000958.1 | - | 5 - 6 | 476 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |