Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PWD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341673.1 | NP_001328878.1 | 32 - 33 | 4026 | 152 | |
NM_001341674.1 | NP_001328879.1 | 32 - 33 | 4030 | 152 | ||
NM_118578.5 | NP_194176.3 | 31 - 32 | 3828 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |