Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118584.3 | NP_194182.1 | 1 - 2 | 681 | 73 | |
GenBank mRNA | AY087262.1 | - | 1 - 2 | 528 | 73 | |
BT026481.1 | - | 1 - 2 | 439 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |