Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203894.1 | NP_001190823.1 | 11 - 12 | 1399 | 76 | |
NM_118585.4 | NP_194183.1 | 11 - 12 | 1474 | 76 | ||
GenBank mRNA | AY054688.1 | - | 11 - 12 | 1367 | 76 | |
BT008426.1 | - | 11 - 12 | 1278 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |