Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBP31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341706.1 | NP_001328008.1 | 4 - 5 | 1126 | 82 | |
NM_118610.4 | NP_194208.1 | 3 - 4 | 1207 | 82 | ||
GenBank mRNA | M94554.1 | - | 3 - 4 | 959 | 82 | |
X65255.1 | - | 971 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |