Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAP24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118621.4 | NP_194219.2 | 1 - 2 | 197 | 126 | |
GenBank mRNA | AY142520.1 | - | 1 - 2 | 181 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |