Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118632.3 | NP_567714.1 | 2 - 3 | 246 | 74 | |
GenBank mRNA | AY065233.1 | - | 2 - 3 | 246 | 74 | |
AY117294.1 | - | 2 - 3 | 226 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |