Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341725.1 | NP_001320060.1 | 7 - 8 | 1613 | 112 | |
NM_118656.5 | NP_194254.2 | 6 - 7 | 1531 | 112 | ||
GenBank mRNA | AY074379.1 | - | 6 - 7 | 1531 | 112 | |
AY091230.1 | - | 6 - 7 | 1394 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |