Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341743.1 | NP_001328956.1 | 7 - 8 | 1291 | 84 | |
NM_001341744.1 | NP_001328957.1 | 8 - 9 | 1197 | 84 | ||
NM_118676.4 | NP_194274.2 | 8 - 9 | 1190 | 84 | ||
GenBank mRNA | AJ252063.1 | - | 7 - 8 | 1054 | 84 | |
BT026122.1 | - | 7 - 8 | 1054 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |