Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036637.2 | NP_001031714.1 | 7 - 8 | 1006 | 91 | |
NM_001084977.1 | NP_001078446.1 | 7 - 8 | 999 | 91 | ||
NM_118677.5 | NP_194275.2 | 7 - 8 | 1120 | 91 | ||
GenBank mRNA | AK175486.1 | - | 7 - 8 | 1004 | 91 | |
AY094470.1 | - | 7 - 8 | 1001 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |