Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DREB1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118680.2 | NP_567720.1 | 1 | 1010 | 73 | |
GenBank mRNA | AB007787.1 | - | 1 | 731 | 73 | |
AF074602.1 | - | 1 | 725 | 73 | ||
EF523095.1 | - | 1 | 731 | 73 | ||
EF523096.1 | - | 1 | 731 | 73 | ||
EF523097.1 | - | 1 | 731 | 73 | ||
EF523098.1 | - | 1 | 731 | 73 | ||
EF523100.1 | - | 1 | 731 | 73 | ||
EF523102.1 | - | 1 | 731 | 73 | ||
EF523103.1 | - | 1 | 731 | 73 | ||
EF523105.1 | - | 1 | 731 | 73 | ||
EF523107.1 | - | 1 | 731 | 73 | ||
EF523114.1 | - | 1 | 731 | 73 | ||
EF523115.1 | - | 1 | 731 | 73 | ||
EF523118.1 | - | 1 | 731 | 73 | ||
EF523122.1 | - | 1 | 731 | 73 | ||
EF523124.1 | - | 1 | 731 | 73 | ||
EF523125.1 | - | 1 | 731 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Inventoried Both primers and probe map within a single exon |