Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118753.3 | NP_194350.1 | 1 - 2 | 293 | 97 | |
GenBank mRNA | AF332390.1 | - | 1 - 2 | 186 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |