Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGP13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118765.3 | NP_194362.1 | 1 | 403 | 151 | |
GenBank mRNA | AF195894.1 | - | 1 | 233 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |