Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VTC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118819.3 | NP_567759.1 | 1 - 2 | 666 | 79 | |
GenBank mRNA | AK226394.1 | - | 1 - 2 | 614 | 79 | |
AY093138.1 | - | 1 - 2 | 611 | 79 | ||
BT006589.1 | - | 1 - 2 | 110 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |