Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT-HF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118824.4 | NP_194420.1 | 14 - 15 | 1405 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |