Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATRBP45C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118834.4 | NP_567764.1 | 4 - 5 | 1121 | 66 | |
GenBank mRNA | AF370339.1 | - | 4 - 5 | 1060 | 66 | |
AY054487.1 | - | 4 - 5 | 1052 | 66 | ||
AY062999.1 | - | 4 - 5 | 969 | 66 | ||
AY093292.1 | - | 4 - 5 | 969 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |