Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203925.2 | NP_001190854.1 | 4 - 5 | 662 | 90 | |
NM_001341872.1 | NP_001320077.1 | 3 - 4 | 1048 | 90 | ||
NM_001341873.1 | NP_001329564.1 | 4 - 5 | 653 | 90 | ||
NM_001341874.1 | NP_001329563.1 | 3 - 4 | 1048 | 90 | ||
NM_001341875.1 | NP_001329565.1 | 4 - 5 | 653 | 90 | ||
NM_118899.4 | NP_194493.2 | 4 - 5 | 811 | 90 | ||
NM_179224.2 | NP_849555.1 | 1 - 2 | 503 | 90 | ||
GenBank mRNA | HM776217.1 | - | 4 - 5 | 539 | 90 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Arabidopsis thaliana Probe spans exons |